EMAIL THIS PAGE TO A FRIEND
LCOR1 Sigma

Long PCR core kit

Purchase

Properties

usage   1 kit sufficient for 125 units polymerase
shipped in   wet ice
storage temp.   −20°C

Description

Caution

It should not be necessary to optimize the Mg++ concentration for KlenTaq LA because of its broad magnesium optimum.

Packaging

Polymerases are not included.

Principle

The long PCR core kit contains all the reagents necessary to perform long PCR on genomic DNA, cDNA, or cloned DNA templates with either AccuTaq LA or KlenTaq LA. Each component has been tested in the Polymerase Chain Reaction.
Optimal concentrations of polymerase, template DNA, primers, and MgCl2 will depend on the specifics of the system. It may be necessary to optimize each component separately, especially the polymerase and Mg++ concentrations and the cycling parameters.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?