• USA Home
  • M0190 - Sartorius Midi Plus pipetting controller
EMAIL THIS PAGE TO A FRIEND
M0190 Sigma
Sartorius Midi Plus pipetting controller
AC/DC input 120 V
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Manufacturer Browser, Pipette Controllers and Pumps, Sartorius Biohit,
mfr. no.   Sartorius 710929
availability   not available in EU
volume   1-100 mL
Description
General description
Biohit Midi Plus of the Biohitfamily is a lightweight cordless pipetting controller for all commonly used glass or plastic pipettes, but can also be used with the Biohit Proline 5 mL and 10 mL disposable tips. The speed can be fine-tuned by applying varying finger pressure to the operating buttons. The Midi Plus can be suspended using a unique spring loaded accessory. When pipetting the Midi Plus can be pulled down to an ideal working height, held in place by tension and locked in position. This optional accessory is ideal for reducing the strain associated with long operating periods. Features include: precise fingertip control of variable speed ranges; hydrophobic autoclavable filter prevents overaspiration; linear speed control; low battery warning.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?