• USA Home
  • M0286 - Ultrafree-MC microcentrifuge filters
EMAIL THIS PAGE TO A FRIEND
M0286 Sigma
Ultrafree-MC microcentrifuge filters
MWCO 5,000 Da, PLCC cellulosic (regenerated cellulose) membrane
Purchase
Properties
Related Categories Centrifuge Filtration, Filtration Supplies, Labware, Manufacturer Browser, Millipore,
material   PLCC cellulosic (regenerated cellulose) membrane
mfr. no.   Millipore, UFC3LCC25
pore size   5,000 Da MWCO
Description
Compatibility
Use in any standard fixed-angle microcentrifuge rotor.
General description
Units consist of microcentrifuge tube with hinged cap and filter insert. The Ultrafree centrifugal filter units are are well suited for the removal of particles & precipitates from aqueous & some solvent-based samples.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?