• USA Home
  • M0313 - [Deamino-Cys3, Nle4, Arg5, β-(2-Naphthyl)-D-Ala7, Cys11]-α-Melanocyte Stimulating Hormone Amide Fragment 3-11

EMAIL THIS PAGE TO A FRIEND
M0313 Sigma

[Deamino-Cys3, Nle4, Arg5, β-(2-Naphthyl)-D-Ala7, Cys11]-α-Melanocyte Stimulating Hormone Amide Fragment 3-11

≥97% (HPLC), solid

Synonym: JKC-366

  • Empirical Formula (Hill Notation) C56H76N18O9S2

  • Molecular Weight 1209.45

  •  MDL number MFCD28016471

  •  PubChem Substance ID 329817769

Purchase

Description

Amino Acid Sequence

Mpa-Nle-Arg-His-Nal-Arg-Trp-Gly-Cys-NH2

Price and Availability


Better Together

Discover your Target

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?