• USA Home
  • M0421 - Met-Lys-[Ser2, Arg3, Pro5, Arg8]-Bradykinin

EMAIL THIS PAGE TO A FRIEND
M0421 Sigma

Met-Lys-[Ser2, Arg3, Pro5, Arg8]-Bradykinin

≥97% (HPLC)

Synonym: Met-Lys-Arg-Ser-Arg-Gly-Pro-Ser-Pro-Arg-Arg

  • CAS Number 165174-60-7

  • Empirical Formula (Hill Notation) C53H98N24O14S

  • Molecular Weight 1327.56

  •  MDL number MFCD00238087

Purchase

Properties

Related Categories Bradykinins, Cell Biology, Cell Signaling and Neuroscience, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   XXSQKDSFQHDKRK-YRYMBYOHSA-N
assay   ≥97% (HPLC)
composition   Peptide content, ~65%
storage temp.   −20°C

Description

Amino Acid Sequence

Met-Lys-Arg-Ser-Arg-Gly-Pro-Ser-Pro-Arg-Arg

Other Notes

A neuropeptide found in Aplysia californica with high sequence homology to bradykinin and kallidin.

Packaging

1 mg in glass bottle

Biochem/physiol Actions

Met-Lys-[Ser2, Arg3, Pro5, Arg8]-Bradykinin (MKRSRGPSPRR) is a neuropeptide found in Aplysia californica with high sequence homology to bradykinin and kallidin. It may be used as a substrate to study the specificity and kinetics of angiotensin-converting enzyme(s) (ANCE).

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?