• USA Home
  • M0536 - Ultrafree-MC microcentrifuge filters
EMAIL THIS PAGE TO A FRIEND
M0536 Sigma
Ultrafree-MC microcentrifuge filters
capacity 400 μL, regenerated cellulose membrane, MWCO 30,000 Da
Purchase
Properties
Related Categories Centrifuge Filtration, Filtration Supplies, Labware, Manufacturer Browser, Millipore,
material   regenerated cellulose membrane
mfr. no.   Millipore UFC3LTK00
  Millipore, UFC3LTK00
  Millipore, UFC3LTK25
  Millipore, UFC3LTK25
limit   5,000 × g max. RCF
capacity   400 μL
pore size   30,000 Da MWCO
Description
Compatibility
Use in any standard fixed-angle microcentrifuge rotor.
General description
Units consist of microcentrifuge tube with hinged cap and filter insert. The Ultrafree centrifugal filter units are are well suited for the removal of particles & precipitates from aqueous & some solvent-based samples.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z677892 Amicon Ultra 0.5 mL centrifugal filters, MWCO 30 kDa
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?