• USA Home
  • M0558 - Whatman® UNIPLATE microplates

EMAIL THIS PAGE TO A FRIEND
M0558 Sigma

Whatman® UNIPLATE microplates

96 well, "V" bottom, well volume 250 μL

Synonym: 96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Purchase

Properties

Related Categories 96 Well Plates, Labware, Manufacturer Browser, Multiwell Plates, Whatman,
material   clear polystyrene plate
  clear polystyrene wells conical bottom
sterility   non-sterile
packaging   pkg of 10 ea
  pkg of 50 ea
mfr. no.   Whatman Article No., 28420880 (US reference)
  Whatman, 7701-6250
size   96 wells
well volume   250 μL
  250 μL
color   clear
matrix active group   Non-treated surface treatment

Description

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

General description

Whatman offers a wide range of UNIPLATE microplates including various well profiles, well volumes, and well densities, in diverse polymer materials. Most UNIPLATE microplates conform to the ANSI/SBS microplate standard and fit most microplate readers and automated plate handling devices.

Whatman UNIPLATE microplates are suitable for a wide range of applications, including simple filtrate collection, when used in conjunction with our UNIFILTER microplates, as well as homogeneous assay techniques utilized in HTS.

Features and Benefits:
• Widest selection from a single source. choice of well volumes ranging from 80 μL to 10 mL, well densities from 24 to 384 wells with round or ′V′ bottom for maximum recovery
• Chemical compatibility
• Available in chemically resistant polymers capable of withstanding low temperatures for long-term storage
• Opaque plates prevent optical cross talk in light emitting assays
• Conforms to ANSI/SBS microplate standard
• Guaranteed for use with robotic handlers and centrifuge carriers

Applications:
• Sample storage
• Assay development
• High-throughput screening
• Plasmid miniprep
• ELISA assays
• Luminescence/chemiluminescence
• Cell culture
• Filtrate collection

Legal Information

Whatman is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?