• USA Home
  • M0683 - Whatman® Multi-Chem 96 well microplates
EMAIL THIS PAGE TO A FRIEND
M0683 Sigma
Whatman® Multi-Chem 96 well microplates
96 well uniplate, 750uL, Multi-Chem, round well bottom, 10/pk
Synonym: 96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates
Purchase
Properties
Related Categories 96 Well Plates, Labware, Multiwell Plates, Whatman 96-Well Multiwell Plates
packaging   pack of 10
mfr. no.   Whatman Article No., 28420881 (US reference)
  Whatman, 7701-6750
availability   not available in EU
size   96 wells
volume   750 μL
Description
General description
Whatman® Multi-Chem 96 Well Microplates
Multi-Chem is a chemically resistant material that exhibits extremely useful properties over a wide range of applications. Providing an excellent choice for storage applications, Multi-Chem microplates are ideal for aggressive organic solvents such as DMF, TFA, THF, acetonitrile, chloroform and methylene chloride. Non-binding properties of Multi-Chem microplates also make them ideal for storage of biological materials.
• Chemically resistant material
• Ideal for aggressive organic solvents
• Non-binding for storage of biological materials
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Multi-Chem is a trademark of Whatman International Ltd.
Whatman is a registered trademark of GE Healthcare
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?