• USA Home
  • M0775 - Methyl 3-O-(N-acetyl-β-D-glucosaminyl)-β-D-galactopyranoside

EMAIL THIS PAGE TO A FRIEND
M0775 Sigma

Methyl 3-O-(N-acetyl-β-D-glucosaminyl)-β-D-galactopyranoside

Synonym: β-D-GlcNAc-(1→3)-β-D-Gal-1→OMe

  • CAS Number 93253-17-9

  • Empirical Formula (Hill Notation) C16H27NO11

  • Molecular Weight 409.39

  •  MDL number MFCD00070380

  •  PubChem Substance ID 24896569

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates M-O, Disaccharide More...
InChI Key   USJPBCYUZSGJII-UHFFFAOYSA-N
storage temp.   −20°C

Description

Application

Used for the inhibition studies of rat lung lectin, Streptococcus pneumoniae, and H. influenzae.

Substrates

Substrate for β-6-GlcNAc-transferase.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?