• USA Home
  • M0911 - Ultrafree-MC microcentrifuge filters
EMAIL THIS PAGE TO A FRIEND
M0911 Sigma
Ultrafree-MC microcentrifuge filters
MWCO 10,000 Da, PTGC polysulfone membrane
Purchase
Properties
Related Categories Centrifuge Filtration, Filtration Supplies, Labware, Manufacturer Browser, Millipore,
material   PTGC polysulfone membrane
mfr. no.   Millipore, UFC3TGC00
  Millipore, UFC3TGC00
pore size   10,000 Da MWCO
Description
Compatibility
Use in any standard fixed-angle microcentrifuge rotor.
General description
Units consist of microcentrifuge tube with hinged cap and filter insert. The Ultrafree centrifugal filter units are are well suited for the removal of particles & precipitates from aqueous & some solvent-based samples.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?