• USA Home
  • M0936 - Greiner 96 well plates, polypropylene

EMAIL THIS PAGE TO A FRIEND
M0936 Sigma

Greiner 96 well plates, polypropylene

2.4 mL, clear polypropylene wells V-bottom

Synonym: 96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Purchase

Properties

Related Categories 96 Well Plates, Greiner, Greiner 96-Well Multiwell Plates, Greiner Multiwell Plates, Greiner Polypropylene 96-Well Plates,
material   polypropylene
  clear polypropylene wells V-bottom
sterility   non-sterile
packaging   case of 50
mfr. no.   Greiner 780270
maximum volume   2,42 mL
size   96 wells
working volume   2.3 mL
color   clear colorless plate
matrix active group   non-treated surface treatment

Description

General description

MICROLON®, 96W Storage Microplate, -20C storage well maximum volume 2 mL. Total well volume 2.42 mL.

Multiwell plates from Greiner. Standard microplate footprint. Excellent optical properties. DNAse/RNAse free. Non-pyrogenic by LAL method to <0.06 EU/mL. Appropriate for applications requiring increased chemical resistance or heat stability, e.g. liquid storage or compound libraries in drug screening.

Other Notes

96 well conical (V) solid bottom microplate, no lid, alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; height 44 mm.

Suitability

Microplates are biologically inert, exhibit low binding characteristics, offer a high temperature tolerance (-196 to -121°C/-321 to -250°F), and are resistant to many standard laboratory chemicals. These properties provide optimal conditions for laboratory specimen and compound storage. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

MICROLON is a registered trademark of Greiner Bio-One GmbH

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?