• USA Home
  • M1051 - 6-Methoxy-1-propyl-1,2,3,4-tetrahydroquinoline

EMAIL THIS PAGE TO A FRIEND
M1051 Sigma

6-Methoxy-1-propyl-1,2,3,4-tetrahydroquinoline

Purchase

Properties

storage temp.   2-8°C

Description

Other Notes

plant material prepared for rbi; not for the Sigma catalog per roy winkel

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?