• USA Home
  • M106 - Mecamylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
M106 Sigma

Mecamylamine hydrochloride

Synonym: 2-(Methylamino)isocamphane hydrochloride, Inversine, N,2,3,3-Tetramethylbicyclo[2.2.1]heptan-2-amine hydrochloride

Purchase

Properties

InChI Key   PKVZBNCYEICAQP-CIISUUNXSA-N
form   neat
format   neat

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
RB6900000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M9020 Mecamylamine hydrochloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?