• USA Home
  • M1145 - Methylphenidate hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
M1145 Sigma

Methylphenidate hydrochloride solution

1.0 mg/mL in methanol

Synonym: threo-Methyl α-phenyl-α-(2-piperidyl)acetate hydrochloride solution, Ritalin hydrochloride

  • Empirical Formula (Hill Notation) C14H19NO2 · HCl

  • Molecular Weight 269.77

Purchase

Properties

drug control   USDEA Schedule II; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Methylphenidate (Ritalin) Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-DA

From our library of Articles, Sigma-Aldrich presents GC Analysis of Methylphenidate (Ritalin) Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-DA
Keywords: Chromatography, Clinical, Flame ionization detector, Forensic, Gas chromatography, Pharmaceutical, Purification

HPLC Analysis of Ritalinic Acid and Ritalin (Methylphenidate) Enantiomers on Astec® CHIROBIOTIC® V2 with T2 Guard Column

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Ritalinic Acid and Ritalin (Methylphenidate) Enantiomers on Astec® CHIROBIOTIC® V2 with T2 Guard Column
Keywords: Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?