• USA Home
  • M1146 - Molybdenum atomic absorption standard solution

EMAIL THIS PAGE TO A FRIEND
M1146 Sigma

Molybdenum atomic absorption standard solution

Purchase

Properties

application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?