• USA Home
  • M1161 - Ultrafree-MC microcentrifuge filters
EMAIL THIS PAGE TO A FRIEND
M1161 Sigma
Ultrafree-MC microcentrifuge filters
capacity 400 μL, polyethersulfone membrane, MWCO 100,000 Da
Purchase
Properties
Related Categories Centrifuge Filtration, Filtration Supplies, Labware, Manufacturer Browser, Millipore,
material   polyethersulfone membrane
mfr. no.   Millipore UFC3BHK00
  Millipore, UFC3BHK00
parameter   5,000 × g max. RCF
capacity   400 μL
pore size   100,000 Da MWCO
Description
Compatibility
Use in any standard fixed-angle microcentrifuge rotor.
General description
Units consist of microcentrifuge tube with hinged cap and filter insert. The Ultrafree centrifugal filter units are are well suited for the removal of particles & precipitates from aqueous & some solvent-based samples.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
CLS431481 Corning® Spin-X® UF concentrators, Spin-X UF 500 for samples up to 500uL; 100K MWCO
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?