EMAIL THIS PAGE TO A FRIEND
M1189 Sigma

Methiazole

98% (TLC)

Synonym: Methyl 5-(isopropylthio)benzimidazole-2-carbamate

  • Empirical Formula (Hill Notation) C12H15N3O2S

  • Molecular Weight 265.33

  •  MDL number MFCD11044449

  •  PubChem Substance ID 24896624

Purchase

Properties

InChI Key   SXJIZQPZESTWLD-UHFFFAOYSA-N
assay   98% (TLC)
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

General description

Broad spectrum anthelmintic with efficiency equal to albendazole. Efficient in cases of polyinvasion, especially against Echinococcus.

White crystalline powder, sometimes grey-shaded.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?