• USA Home
  • M1256 - 6-Methylpurine riboside

EMAIL THIS PAGE TO A FRIEND
M1256 Sigma

6-Methylpurine riboside

Synonym: 6-Methyl-9-(1-β-D-ribofuranosyl)purine

  • CAS Number 14675-48-0

  • Empirical Formula (Hill Notation) C11H14N4O4

  • Molecular Weight 266.25

  •  PubChem Substance ID 329817864

Purchase

Properties

InChI Key   FIGBCBGMUIGJBD-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?