EMAIL THIS PAGE TO A FRIEND
M1285 Sigma

M16 medium

mouse embryo tested, powder

Purchase

Properties

Quality Level   GMP
form   powder
suitability   mouse embryo tested
storage temp.   2-8°C

Description

Application

Recommended for embryo culture in a CO2 incubator.

Quantity

Formulated to contain 11.5 grams of powder per liter of medium.

Reconstitution

Supplement with 2.010 g/L sodium bicarbonate, 0.06 g/L potassium penicillin-G, 0.05 g/L streptomycin sulfate, and 4.349 DL-lactic acid (sodium). See product insert for details.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M7292 M16 Medium, With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?