EMAIL THIS PAGE TO A FRIEND
M1399 Sigma

Sodium fluoroacetate

~95%

Synonym: Fluoroacetic acid sodium salt, Gifblaar poison

Purchase

Properties

InChI Key   JGFYQVQAXANWJU-UHFFFAOYSA-M
assay   ~95%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2629 6.1 / PGI
WGK Germany 
3
RTECS 
AH9100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?