• USA Home
  • M1414 - Mycoplasma Control Slides

EMAIL THIS PAGE TO A FRIEND
M1414 Sigma

Mycoplasma Control Slides

Purchase

Properties

Related Categories Cell Culture, Mycoplasma Detection, Reagents and Supplements More...
Quality Level   GMP
storage temp.   2-8°C

Description

Application

For use in Mycoplasma stain kit.

Other Notes

Cells are grown on coverslips which have been mounted on microscope slides. Use caution when handling control slides as cells are exposed to facilitate staining.

Price and Availability


All labs need water

Standard component

Description

Product #

Add to Cart

negative control slides 10 SDS    
positive control slides 10 SDS    
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Mycoplasma Detection/Elimination

The maintenance of contamination-free cell lines is essential to cell-based research. Among the biggest concerns are mycoplasma contamination. Although mycoplasma do not usually kill contaminated cel...
Don Finley
BioFiles v7 n1, 2011, 14–15
Keywords: Eliminations, Metabolism, Polymerase chain reaction

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?