• USA Home
  • M1677 - Matrix Metalloproteinase-3 from human fibroblasts

EMAIL THIS PAGE TO A FRIEND
M1677 Sigma

Matrix Metalloproteinase-3 from human fibroblasts

≥95% (SDS-PAGE), buffered aqueous solution

Synonym: MMP-3, Proteoglycanase, Stromelysin-1, Transin

Purchase

Description

Biochem/physiol Actions

Breaks down proteoglycan, fibronectin, laminin, and type IV collagen, but not interstitial type I collagen.

Physical form

Solution in 50 mM Tris, pH 7.4, 0.5 M sodium chloride, 10 mM calcium chloride, 0.05% Brij-35, and 0.02% sodium azide.

Price and Availability

Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?