• USA Home
  • M1802 - Matrix Metalloproteinase-1 human

EMAIL THIS PAGE TO A FRIEND
M1802 Sigma

Matrix Metalloproteinase-1 human

≥90% (SDS-PAGE), recombinant, expressed in transfected cells, buffered aqueous solution

Synonym: Collagenase-1, Interstitial Collagenase, MMP-1

Purchase

Description

Biochem/physiol Actions

Initiates the breakdown of interstitial collagens.

MMP-1 initiates the breakdown of interstitial collagens.

Physical form

Supplied as a solution in 50 mM Tris HCl, pH 7.0, containing 150 mM sodium chloride, 10 mM calcium chloride, and 1 mM magnesium chloride.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

M9195 Matrix Metalloproteinase-1 human, recombinant, expressed in NSO cells, >95% (SDS-PAGE), buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?