• USA Home
  • M1927 - Matrix Metalloproteinase-2 from human fibroblasts

EMAIL THIS PAGE TO A FRIEND
M1927 Sigma

Matrix Metalloproteinase-2 from human fibroblasts

>95% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: 72 Kd Gelatinase, 72 Kd Type IV collagenase, Gelatinase A, MMP-2

Purchase

Description

Biochem/physiol Actions

Degrades general matrix components and may have a role in processes such as host defense, cell proliferation, and protein turnover as well as tissue remodeling.

Quality

The enzyme is free of its endogenous inhibitor, TIMP-2 and other matrix metalloproteinases.

Specificity

Specifically cleaves type IV collagen, a major structural component of basement membranes.

Physical form

Solution in 10 mM sodium phosphate, pH 7.4, 250 mM sodium chloride, 2% DMSO, and 50% (v/v) glycerol.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
1
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

M9070 Matrix Metalloproteinase-2 human, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?