EMAIL THIS PAGE TO A FRIEND
M1928 Sigma

MYP agar base

BioChemika, for microbiology

Synonym: Mannitol Egg Yolk Polymyxin Agar Base

Purchase

Description

Components

Ingredients (g/L)
Peptic digest of animal tissue, 10.00
Meat extract, 1.00
D-Mannitol, 10.00
Sodium chloride, 10.00
Phenol red, 0.025
Agar, 15.00

Application

Used for isolation and identification of Bacillus and Staphylococci.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

EZ-Product Family for Bioburden Testing

The EZ family of products has been designed to optimize and streamline the bioburden workflow. The family includes:
Keywords: Filtration, Separation

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

P9602 Polymyxin B selective supplement, for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?