• USA Home
  • M194 - M1 Muscarinic Receptor human

EMAIL THIS PAGE TO A FRIEND
M194 Sigma

M1 Muscarinic Receptor human

recombinant, expressed in CHO cells, membrane suspension

Purchase

Properties

recombinant   expressed in CHO cells
form   membrane suspension
packaging   vial of 50 units
storage temp.   −70°C

Description

Analysis Note

Recommended assay conditions are supplied on the product data sheet. This is a bioassayed product.

Physical form

Membrane suspension in 10 mM Tris-HCl at pH 7.2, containing 2 mM EDTA. Actual concentrations and specific binding capacity are provided with each lot

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?