• USA Home
  • M2010 - Molecular sieves, 3 Å

EMAIL THIS PAGE TO A FRIEND
M2010 Sigma

Molecular sieves, 3 Å

1/8 in. (pellet)

Synonym: Molecular Sieve UOP Type 3Å

Purchase

Properties

particle size   1/8 in. (pellet)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?