EMAIL THIS PAGE TO A FRIEND
M2053 Sigma

MN Agar

for microbiology

Purchase

Properties

Related Categories All Solid Media/Agars, Media, Microbiology More...
grade   for microbiology

Description

Components

Ingredients (g/L)
Beef extract, 1.00
Yeast extract, 0.075
Manganous carbonate, 2.00
Ferrous ammonium sulfate, 0.15
Sodium citrate, 0.15
Agar, 12.00

Application

Used for detection of Leptothrix by its ability to oxidize the Mn2+ ion.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

C3607 Cyanocobalamin, meets USP testing specifications

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?