• USA Home
  • M2059 - Multiwash Advantage®

EMAIL THIS PAGE TO A FRIEND
M2059 Sigma

Multiwash Advantage®

8-port Coaxial Manifold

Purchase

Properties

Related Categories Labware, Manufacturer Browser, Multiwell Plates, Tricontinent, Washers, Orienters, etc. More...
sterility   non-sterile
mfr. no.   Tricontinent
color   none
matrix active group   Non-treated surface treatment

Description

General description

Engineered for reliability, with features like the QuickStart Guide and quick-connect fittings that speed it into full operation in less than 15 minutes, this has the versatility of easy-to-program wash routines, bottom washing, sweeping crosswise aspiration, and overflow washing capabilities that allow it to perform tests that include:
cell washing; ELISA; agglutination tests; EIA, FIA, and LIA; cytotoxicity tests.
The MultiWash Advantage performs a 300 mL three-cycle wash on a full 96 well plate in less than two minutes. By programming and storing routines, technicians can switch
between seven different modes at the touch of a button. Additional features include a floating manifold that adjusts to match each plate and allows the washer to perform special test procedures. At the end of the day, the MultiWash Advantage provides a push button rinse-on-demand feature to clean the system.

Legal Information

Advantage is a registered trademark of Mine Safety Appliances Co.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?