EMAIL THIS PAGE TO A FRIEND
M2063 Sigma

5-Maleimido-eosin

Synonym: Eosin-5-maleimide

  • CAS Number 150322-02-4

  • Empirical Formula (Hill Notation) C24H9Br4NO7

  • Molecular Weight 742.95

  •  MDL number MFCD07437936

Purchase

Properties

InChI Key   YRCLWXRIEYQTAV-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?