• USA Home
  • M2161 - Ultrafree-CL microcentrifuge filters
EMAIL THIS PAGE TO A FRIEND
M2161 Sigma
Ultrafree-CL microcentrifuge filters
MWCO 30,000 Da, PTTK polysulfone membrane
Purchase
Properties
Related Categories Centrifuge Filtration, Filtration Supplies, Labware, Manufacturer Browser, Millipore,
material   PTTK polysulfone membrane
feature   RCF≤ 5,000 × g
mfr. no.   Millipore, UFC4TTK25
capacity   2.0 mL
pore size   30,000 Da MWCO
Description
Compatibility
Use in any standard fixed-angle rotor that accepts 15 mL or 20 mm O.D. centrifuge tubes.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z648035 Amicon Ultra centrifugal filter units, Ultra-4, MWCO 30 kDa
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?