• USA Home
  • M2256 - N-Methyl-N-nitroso-p-toluenesulfonamide

EMAIL THIS PAGE TO A FRIEND
M2256 Sigma

N-Methyl-N-nitroso-p-toluenesulfonamide

Synonym: p-Tolylsulfomethylnitrosamide, Diazald®, Diazomethane precursor, N-Methyl-N-(p-tolylsulfonyl)nitrosamide, N-Nitroso-p-toluenesulfomethylamide, N-Nitroso-N-methyl-p-toluenesulfonamide

  • CAS Number 80-11-5

  • Empirical Formula (Hill Notation) C8H10N2O3S

  • Molecular Weight 214.24

  •  Beilstein Registry Number 2214345

  •  EC Number 201-252-6

  •  MDL number MFCD00002050

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   FFKZOUIEAHOBHW-UHFFFAOYSA-N

Description

Application

Reagent for generating diazomethane.

Legal Information

Diazald is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3234 4.1/PG 2
WGK Germany 
2
RTECS 
XT5950000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

D28000 Diazald®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?