• USA Home
  • M226 - p-MPPF dihydrochloride

EMAIL THIS PAGE TO A FRIEND
M226 Sigma

p-MPPF dihydrochloride

solid

Synonym: 4-Fluoro-N-(2-[4-(2-methoxyphenyl)1-piperazinyl]ethyl)-N-(2-pyridinyl)benzamide dihydrochloride

  • Empirical Formula (Hill Notation) C25H27FN4O2 · 2HCl

  • Molecular Weight 507.43

  •  MDL number MFCD01321045

  •  PubChem Substance ID 24278581

Purchase

Properties

Related Categories Antagonists, Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, GPCR Modulators,
InChI Key   TZOICBMBENXHNK-UHFFFAOYSA-N
form   solid
color   white
solubility   ethanol: soluble15 mg/mL
  H2O: soluble19 mg/mL
storage temp.   2-8°C
Gene Information   human ... HTR1A(3350)

Description

Biochem/physiol Actions

Selective 5-HT1A serotonin receptor antagonist. More potent than p-MPPI.

Caution

Hygroscopic

Legal Information

Manufactured and sold under license from the University of Pennsylvania, U.S. Patent 5,744,121.

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?