• USA Home
  • M231 - 3-Methoxynaltrexone hydrochloride

EMAIL THIS PAGE TO A FRIEND
M231 Sigma

3-Methoxynaltrexone hydrochloride

solid

  • Empirical Formula (Hill Notation) C21H25NO4 · HCl

  • Molecular Weight 391.89

  •  MDL number MFCD01530048

  •  PubChem Substance ID 329817937

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Excitatory Amino Acids - Opiod Antagonists, GPCR Modulators,
InChI Key   BWFKBOSEDNYSHX-OPHZJPRHSA-N
form   solid
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
color   white
solubility   H2O: 22 mg/mL

Description

Caution

Photosensitive

Biochem/physiol Actions

Putative antagonist at a novel opioid receptor that binds morphine-6-β-glucuronide; blocks endomorphin-2-induced antinociception.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?