• USA Home
  • M2422 - Maleic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
M2422 Sigma

Maleic acid sodium salt

BioXtra, ≥99%

Synonym: cis-Butenedioic acid, Toxilic acid

Purchase

Properties

Related Categories Biochemicals and Reagents, Dicarboxylic acids, Fatty Acids and conjugates, Fatty Acyls, Lipids,
InChI Key   VRVKOZSIJXBAJG-ODZAUARKSA-M
product line   BioXtra
assay   ≥99%
impurities   <0.0005% Phosphorus (P)
  <0.1% Insoluble matter
solubility   H2O: soluble1 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): <0.05%
  sulfate (SO42-): <0.05%
cation traces   Al: <0.0005%
  Ca: <0.0005%
  Cu: <0.0005%
  Fe: <0.0005%
  K: <0.005%
  Mg: <0.0005%
  NH4+: <0.05%
  Pb: <0.001%
  Zn: <0.0005%

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?