• USA Home
  • M2433 - Media bottles, wide mouth, Lab 45
EMAIL THIS PAGE TO A FRIEND
M2433 Sigma
Media bottles, wide mouth, Lab 45
size 100 mL
Synonym: bottle wide mouth, wide mouth bottle, wide mouth bottles
Purchase
Properties
Related Categories Bottles and Caps, Glass Media Bottles, Labware, Media Bottles
description   Autoclavable
mfr. no.   Wheaton Scientific
size   100 mL
Description
General description
Graduated safety coated bottles are ideal for storing fluids, dry chemicals, and other materials in the lab and help protect against broken glass. The strong, slip resistant plastisol coating reduces potential breakage and exposure to lab personnel handling hazardous material. Manufactured from Wheaton glass (meets USP Type I standards). Fluids are protected because the glass has a low alkali content. Wide opening, special lip and sloping shoulders facilitate pouring and cleaning. Includes a white polypropylene screw cap with a wedge-shaped inner sealing ring and polypropylene pour ring.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
recommended
Product #
Description
Add to Cart
C1351 Media bottles, wide mouth, Lab 45, accessory, PTFE-lined phenolic caps
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?