• USA Home
  • M2436 - Greiner 96 well plates, polystyrene

EMAIL THIS PAGE TO A FRIEND
M2436 Sigma

Greiner 96 well plates, polystyrene

clear polystyrene wells round bottom, sterile

Synonym: 96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Purchase

Properties

Related Categories 96 Well Plates, Greiner, Greiner 96-Well Multiwell Plates, Greiner Multiwell Plates, Greiner Polystyrene 96-Well Plates,
material   clear polystyrene plate
  clear polystyrene wells round bottom
sterility   sterile
feature   lid: no
packaging   case of 100
  pkg of (packs of 5)
mfr. no.   Greiner 650163
maximum volume   323 μL
size   96 wells
working volume   280 μL
color   clear
matrix active group   non-treated surface treatment

Description

General description

96W Microplate, well working volume 40-280 μL; maximum volume 323 μL. Growth surface per well 0.35 cm(2).

Greiner multiwell plates are available in the standard multiwell plate footprint and offer excellent optical properties. These plates are DNAse/RNAse free and non-pyrogenic as deteremined by LAL method (< 0.06 EU/mL). Appropriate for general purposes, such as optical measurements, photography, and microscopy, due to the high transparency of polystyrene plates.

Other Notes

96 well microplate, without lid, solid round (U) bottom well design, alphanumeric well coding, length: 127.76+/- 0.15 mm width: 85.48 +/- 0.15 mm curvature: approx.200 μm.

Suitability

Suitable for use with live cells. Free of detectable DNase/RNase, human DNA and pyrogens.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?