EMAIL THIS PAGE TO A FRIEND
M2439 Sigma

Maltotetraitol

≥97%, solid

  • Empirical Formula (Hill Notation) C24H44O21

  • Molecular Weight 668.59

  •  MDL number MFCD00083651

  •  PubChem Substance ID 329817941

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates M-O, Polysaccharide More...
InChI Key   LUAHEUHBAZYUOI-KVXMBEGHSA-N
assay   ≥97%
form   solid
storage temp.   2-8°C

Description

General description

Maltotetraitol is a sugar alcohol.

Application

Maltotetraitol has been used in a study to assess two modes of ligand binding in the maltose-binding protein of Escherichia coli. It has also been used in a study to investigate the effects of reduced malto-oligosaccharides on the thermal stability of pullulanase from Bacillus acidopullulyticus.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?