• USA Home
  • M251 - Macrophage-Derived Chemokine human

EMAIL THIS PAGE TO A FRIEND
M251 Sigma

Macrophage-Derived Chemokine human

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, cell culture tested

Synonym: MDC, STCP-1

Purchase

Properties

Related Categories Cell Biology, Cell Culture, Cell Signaling and Neuroscience, Chemokines, Cytokines and Growth Factors,
recombinant   expressed in E. coli
assay   >97% (SDS-PAGE)
form   lyophilized powder
impurities   endotoxin, tested
suitability   cell culture tested
storage temp.   −20°C
Gene Information   human ... CCL22(6367)

Description

Analysis Note

The biological activity is measured by its ability to chemoattract human T-lymphoblastoid CEM-NKr cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in 20% acetonitrile and 0.1% trifluoroacetic acid containing 1.25 mg bovine serum albumin.

Biochem/physiol Actions

Macrophage-Derived Chemokine (MDC) induces chemotaxis or calcium mobilization in dendritic cells, IL-2 activated cells, and activated T lymphocytes. MDC, also named stimulated T cell chemotactic protein (STCP-1), shows <35% identity to other CC chemokine family members. It is expressed in dendritic cells, macrophages, and activated monocytes. Expression is also detected in the tissues of thymus, lymph node, and appendix.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?