EMAIL THIS PAGE TO A FRIEND
M2543 Sigma

Mesityl oxide

suitable for neutral marker for measuring electroosmotic flow (EOF), ~98%

Synonym: 4-Methyl-3-penten-2-one

Purchase

Properties

InChI Key   SHOJXDKTYKFBRD-UHFFFAOYSA-N
assay   ~98%
refractive index   n20/D 1.442(lit.)
bp   130 °C(lit.)
mp   -41.5 °C(lit.)
density   0.858 g/mL at 25 °C(lit.)
suitability   suitable for neutral marker for measuring electroosmotic flow (EOF)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 1229 3 / PGIII
WGK Germany 
1
RTECS 
SB4200000
Flash Point(F) 
87.8 °F
Flash Point(C) 
31 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?