• USA Home
  • M2556 - Anti-Matrix Metalloproteinase-11, Propeptide Region antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
M2556 Sigma

Anti-Matrix Metalloproteinase-11, Propeptide Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-MMP-11

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Extracellular Matrix, Antibodies to Matrix Metalloproteinases and TIMPs,
species reactivity   human
application(s)   immunohistochemistry (frozen sections): suitable
  immunoprecipitation: suitable
  indirect ELISA: suitable
  western blot: 1:1,000
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... MMP11(4320)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the propeptide region of human matrix metalloproteinase-11 (stromelysin-3).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide.

Specificity

Reacts with reduced and non-reduced MMP-11. Recognizes the pro-form of MMP-11 at 58 kDa but not the active form at 45 kDa.

Price and Availability


Antibody Explorer

Explore with Confidence

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany  1
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?