• USA Home
  • M2636 - Methyl 2,3-O-isopropylidene-5-O-(p-tolylsulfonyl)-β-D-ribofuranoside

EMAIL THIS PAGE TO A FRIEND
M2636 Sigma

Methyl 2,3-O-isopropylidene-5-O-(p-tolylsulfonyl)-β-D-ribofuranoside

≥98% (TLC)

  • CAS Number 4137-56-8

  • Empirical Formula (Hill Notation) C16H22O7S

  • Molecular Weight 358.41

  •  MDL number MFCD00038411

  •  PubChem Substance ID 24896749

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates M-O, Monosaccharide More...
InChI Key   IAPMZKRZMYQZSW-KBUPBQIOSA-N
assay   ≥98% (TLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?