• USA Home
  • M278 - 8-Methoxymethyl-3-isobutyl-1-methylxanthine

EMAIL THIS PAGE TO A FRIEND
M278 Sigma

8-Methoxymethyl-3-isobutyl-1-methylxanthine

solid

Synonym: 3,7-Dihydro-8-(methoxymethyl)-1-methyl-3-(2-methylpropyl)-1H-purine-2,6-dione, 8-Methoxymethyl IBMX

Purchase

Properties

form   solid
color   white
solubility   H2O: 0.3 mg/mL
  DMSO: 4.4 mg/mL

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M2547 8-Methoxymethyl-3-isobutyl-1-methylxanthine, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?