Synonym:
5-
CAS Number 101043-37-2
Empirical Formula (Hill Notation) C49H74N10O12
Molecular Weight 995.17
MDL number MFCD00131607
PubChem Substance ID 24896778
Popular Documents: Datasheet (PDF)
| Related Categories | Application Index, Bacteria, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes, |
| assay | ≥95% (HPLC) |
| form | solid film |
| solubility | ethanol: soluble1 mg/mL |
| DMSO: soluble | |
| storage temp. | −20°C |
Potent inhibitor of protein phosphatase Types 1 and 2A; has no effect on protein kinase. Hepatic tumor promoter in experimental animal model.1
Hepatotoxic cyclic heptapeptide.
Stock solutions can be diluted in 50 mM Tris-HCl, pH 7.0, containing 0.03% Brij 35; phosphatase inhibition will be retained for up to 1 month when stored at concentrations up to 0.01 nM.
Customers Also Viewed
cyanotoxin
5 μg/mL in methanol (each), analytical standard, for environmental analysis
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?