• USA Home
  • M3006 - 2′-O-Monobutyrylguanosine 3′:5′-cyclic monophosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
M3006 Sigma

2′-O-Monobutyrylguanosine 3′:5′-cyclic monophosphate sodium salt

  • CAS Number 58329-72-9

  • Empirical Formula (Hill Notation) C14H17N5O8P · Na

  • Molecular Weight 437.28

  •  MDL number MFCD00057112

  •  PubChem Substance ID 24896610

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotide Analogs More...
InChI Key   SWTMQRFILNAADD-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?