• USA Home
  • M3038 - Methoxypolyethylene glycol tresylate

EMAIL THIS PAGE TO A FRIEND
M3038 Sigma

Methoxypolyethylene glycol tresylate

≥65%, mol wt ~5,000

Synonym: TMPEG

Purchase

Properties

Related Categories Molecular Biology, PEGylation, Protein Modification, Protein Structural Analysis, Proteomics More...
assay   ≥65%
mol wt   mol wt ~5,000
storage temp.   −20°C

Description

General description

Methoxypolyethylene glycol tresylate is a polyethylene glycol (PEG) derivative. PEGs are widely used as a covalent modifier of biologic macromolecules and particulates and for mechanism of drug delivery.

Application

Methoxypolyethylene glycol has been used in a study to assess the effect of PEGylation on Bacteriorhodopsin stability towards ethanol. It has also been used in a study to investigate hydrogel containing silica shell cross-linked micelles for drug delivery. It is also used for modification of proteins under mild conditions.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?