• USA Home
  • M3139 - α-Methyl D-mannoside-agarose

EMAIL THIS PAGE TO A FRIEND
M3139 Sigma

α-Methyl D-mannoside-agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Carbohydrate Matrices, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
matrix   6% beaded agarose
matrix activation   divinyl sulfone
matrix attachment   hydroxyl
matrix spacer   5 atoms
capacity   10-15 mg/mL binding capacity (Concanavalin A)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.15 M NaCl, 0.01 M sodium phosphate, pH 6.8, containing 0.02% sodium azide

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?