• USA Home
  • M3170 - α,β-Methyleneguanosine 5′-diphosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
M3170 Sigma

α,β-Methyleneguanosine 5′-diphosphate sodium salt

≥98%

Synonym: GMPCP, GPCP

  • CAS Number 161308-39-0

  • Empirical Formula (Hill Notation) C11H17N5O10P2 · xNa+

  • Molecular Weight 441.23 (free acid basis)

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotide Analogs More...
assay   ≥98%
storage temp.   −20°C

Description

Packaging

25 mg in poly bottle

5 mg in glass bottle

Application

α,β-Methyleneguanosine 5′-diphosphate (GPCP; GMPCP) is a GDP methylene analog used to study vinblastine-induced self-association of tubulin. GMPCP is used as an alternative to calcium to produce helical tubes that support bacterial division via assembly of cytokinetic ring structures, Z-rings. GMPCP may be used in comparison with the more slowly hydrolyzed analog guanylyl-(α,β)-methylene-diphosphonate (GMPCPP).

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?