• USA Home
  • M3266 - Myokinase from Bacillus stearothermophilus

EMAIL THIS PAGE TO A FRIEND
M3266 Sigma

Myokinase from Bacillus stearothermophilus

lyophilized powder, ≥150 units/mg protein

Synonym: ATP:AMP phosphotransferase, Adenylate Kinase

Purchase

Properties

Related Categories Cell Biology, General Metabolic Enzymes, Metabolomics, Nucleotide Metabolism
form   lyophilized powder
composition   Protein, ~50% biuret
storage temp.   2-8°C

Description

Unit Definition

One unit will convert 2.0 μmoles of ADP to ATP + AMP per min at pH 7.6 at 37 °C.

Physical form

Lyophilized powder containing Tris buffer salts, pH 8.5

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

M0196 Myokinase from Bacillus stearothermophilus, lyophilized powder, ≥150 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?