• USA Home
  • M3268 - (±)-Methadone hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
M3268 Sigma

(±)-Methadone hydrochloride solution

drug standard, 1.0 mg/mL in methanol

Purchase

Properties

Related Categories Additional Standards, Alkaloids, Analytical Standards, Analytical/Chromatography, Chemical Structure,
grade   drug standard
InChI Key   FJQXCDYVZAHXNS-UHFFFAOYSA-N
drug control   Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschlandregulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

GC Analysis of Basic Drugs on Equity®-5

From our library of Articles, Sigma-Aldrich presents GC Analysis of Basic Drugs on Equity®-5
Keywords: Chromatography, Gas chromatography, Purification

HPLC Analysis of Methadone Enantiomers and Methadone Metabolite EDDP on Astec® CYCLOBOND® I 2000 HP-RSP

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Methadone Enantiomers and Methadone Metabolite EDDP on Astec® CYCLOBOND® I 2000 HP-RSP
Keywords: Analgesics, Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Methadone Enantiomers on Astec® CHIROBIOTIC® V2

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Methadone Enantiomers on Astec® CHIROBIOTIC® V2
Keywords: Analgesics, Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?